Gene Transcripts

Gene Symbol
Plekha4
Entrez Gene ID
69217
Gene Name
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 4
Gene Aliases
2410005C22Rik, C88169, Pepp1
Chromosome Location
Chr. 7: 45526330 - 45554229 on Build GRCm38
UniGene ID
Mm.274158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148927.2 NP_683729.2 7 729
GenBank mRNA BC021387.1 - 7 740
BC024961.1 - 5 688 4945500

View Full Product Details