Gene Transcripts

Gene Symbol
Slc35f4
Entrez Gene ID
75288
Gene Name
solute carrier family 35, member F4
Gene Aliases
4930550L21Rik
Chromosome Location
Chr. 14: 49298519 - 49525837 on Build GRCm38
UniGene ID
Mm.78738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029238.2 NP_083514.1 7, 8 1441
GenBank mRNA AB041538.1 - 8, 9 1369
AK034308.1 - 7, 8 1443
BC138981.1 - 7, 8 1340 8862003
BC145720.1 - 7, 8 1340 40131023

View Full Product Details