Gene Transcripts
- Gene Symbol
- Pcyt1a
- Entrez Gene ID
- 13026
- Gene Name
- phosphate cytidylyltransferase 1, choline, alpha isoform
- Gene Aliases
- CTalpha, Cctalpha, Ctpct, Cttalpha
- Chromosome Location
- Chr. 16: 32430921 - 32475065 on Build GRCm38
- UniGene ID
- Mm.98775
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163159.1 |
NP_001156631.1
|
4, 5 |
611 |
|
NM_001163160.1 |
NP_001156632.1
|
4, 5 |
545 |
|
NM_009981.4 |
NP_034111.1
|
4, 5 |
447 |
|
GenBank mRNA |
AK044585.1 |
- |
4, 5 |
448 |
|
AK076050.1 |
- |
4, 5 |
372 |
|
AK076830.1 |
- |
4, 5 |
546 |
|
BB642590.1 |
- |
4, 5 |
448 |
|
BC018313.1 |
- |
3, 4 |
330 |
4162053 |
L28956.1 |
- |
4, 5 |
416 |
|
Z12302.1 |
- |
4, 5 |
406 |
|
View Full Product Details