Gene Transcripts

Gene Symbol
Zfy1
Entrez Gene ID
22767
Gene Name
zinc finger protein 1, Y linked
Gene Aliases
Zfy-1
Chromosome Location
Chr. Y: 725207 - 797409 on Build GRCm38
UniGene ID
Mm.4324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009570.4 NP_033596.3 8 1305
GenBank mRNA AK076618.1 - 8 1306
X14382.1 - 8 1232

View Full Product Details