Gene Transcripts
- Gene Symbol
- Acin1
- Entrez Gene ID
- 56215
- Gene Name
- apoptotic chromatin condensation inducer 1
- Gene Aliases
- 2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670
- Chromosome Location
- Chr. 14: 54642161 - 54686931 on Build GRCm38
- UniGene ID
- Mm.297078
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001085472.2 |
NP_001078941.2
|
9, 10 |
947 |
|
NM_001085473.2 |
NP_001078942.2
|
15, 16 |
3272 |
|
NM_001242605.1 |
NP_001229534.1
|
8, 9 |
1011 |
|
NM_001242606.1 |
NP_001229535.1
|
8, 9 |
972 |
|
NM_019567.3 |
NP_062513.3
|
9, 10 |
1076 |
|
NM_023190.3 |
NP_075679.2
|
15, 16 |
3308 |
|
GenBank mRNA |
AF124725.1 |
- |
9, 10 |
1007 |
|
AF124729.1 |
- |
8, 9 |
942 |
|
AF168782.1 |
- |
15, 16 |
3309 |
|
AK012099.1 |
- |
9, 10 |
940 |
|
AK030449.1 |
- |
15, 16 |
3002 |
|
AK122342.1 |
- |
15, 16 |
2943 |
|
BC019127.1 |
- |
8, 9 |
996 |
|
BC052755.1 |
- |
8, 9 |
932 |
|
BC075691.1 |
- |
15, 16 |
2957 |
|
BC094217.1 |
- |
8, 9 |
954 |
|
View Full Product Details