Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025829.4 | NP_080105.1 | 6 | 506 | |
GenBank mRNA | AK005054.2 | - | 6 | 507 | |
AK007268.1 | - | 6 | 495 | ||
AK035794.1 | - | 310 | |||
AY628329.1 | - | 6 | 487 | ||
BC027014.1 | - | 6 | 501 | 5369891 |