Gene Transcripts

Gene Symbol
Rabep2
Entrez Gene ID
70314
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
2610011A08Rik, AI462746, Fra
Chromosome Location
Chr. 7: 126428416 - 126445907 on Build GRCm38
UniGene ID
Mm.35467
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030566.2 NP_085043.2 2, 3 598
GenBank mRNA AK089938.1 - 2, 3 302
AK144836.1 - 2, 3 284
AK157832.1 - 2, 3 259
BC004088.1 - 1, 2 344 3593722
BC015287.1 - 2, 3 606 4240442
BU514356.1 - 2, 3 598

View Full Product Details