Gene Transcripts

Gene Symbol
Oip5
Entrez Gene ID
70645
Gene Name
Opa interacting protein 5
Gene Aliases
5730547N13Rik, Lint-25, Ms18b
Chromosome Location
Chr. 2: 119609532 - 119618505 on Build GRCm38
UniGene ID
Mm.246412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042653.1 NP_001036118.1 3 516
GenBank mRNA AK017825.1 - 3 481
AK045323.1 - 2 719
BC115883.1 - 3 472 40088361
BC116660.1 - 3 473 40088363

View Full Product Details