Gene Transcripts

Gene Symbol
Nhlrc4
Entrez Gene ID
621239
Gene Name
NHL repeat containing 4
Gene Aliases
F430201B04Rik
Chromosome Location
Chr. 17: 25942233 - 25944931 on Build GRCm38
UniGene ID
Mm.381070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039038.2 NP_001034127.1 2 595
GenBank mRNA AK143822.1 - 2 595
BC147284.1 - 1 185 9056317
BC147285.1 - 1 184 9056337

View Full Product Details