Gene Transcripts

Gene Symbol
Stk38
Entrez Gene ID
106504
Gene Name
serine/threonine kinase 38
Gene Aliases
5830476G13Rik, 9530097A09Rik, AA617404, Ndr1
Chromosome Location
Chr. 17: 28970885 - 29008006 on Build GRCm38
UniGene ID
Mm.439986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134115.2 NP_598876.1 4 481
GenBank mRNA AK035731.1 - 4 490
AK080179.1 - 4 490
AY292399.1 - 4 345
BC009658.1 - 4 481 3484764


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK153676.1 - 1422 Chr. - - [-]

View Full Product Details