Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178926.3 | NP_849257.1 | 1, 2 | 293 | |
GenBank mRNA | AK078549.1 | - | 1, 2 | 224 | |
AK089265.1 | - | 1, 2 | 350 | ||
AK145854.1 | - | 1, 2 | 146 | ||
AK170944.1 | - | 1, 2 | 239 | ||
BC026472.1 | - | 1, 2 | 293 | 4487505 |