Gene Transcripts

Gene Symbol
Slc5a10
Entrez Gene ID
109342
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 10
Gene Aliases
C330021F16Rik, SGLT5
Chromosome Location
Chr. 11: 61672782 - 61720811 on Build GRCm38
UniGene ID
Mm.441070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033227.2 NP_001028399.1 5 570
GenBank mRNA AK144033.1 - 5 571
AK165337.1 - 4 366
BC141325.1 - 4 363 9055937

View Full Product Details