Gene Transcripts

Gene Symbol
Zkscan2
Entrez Gene ID
210162
Gene Name
zinc finger with KRAB and SCAN domains 2
Gene Aliases
9430065N20Rik, AW493225, Zfp694
Chromosome Location
Chr. 7: 123477747 - 123504904 on Build GRCm38
UniGene ID
Mm.135397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081329.1 NP_001074798.1 7 2479
GenBank mRNA AK034954.1 - 7 2481
BC028272.1 - 1 205
BC054371.1 - 1 194

View Full Product Details