Gene Transcripts

Gene Symbol
Fmo6
Entrez Gene ID
226565
Gene Name
flavin containing monooxygenase 6
Gene Aliases
Chromosome Location
Chr. 1: 162916551 - 162937572 on Build GRCm38
UniGene ID
Mm.270516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178038.1 NP_001171509.1 6 1226
GenBank mRNA BC051402.1 - 7 1313

View Full Product Details