Gene Transcripts

Gene Symbol
Lsm2
Entrez Gene ID
27756
Gene Name
LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
D17H6S56E-2, D17H6S56E2, Dmapl, Dmpkap, G7b, Sm-X5, SmX5, snRNP
Chromosome Location
Chr. 17: 34981854 - 34985891 on Build GRCm38
UniGene ID
Mm.165735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110101.2 NP_001103571.1 4, 5 380
NM_001204273.1 NP_001191202.1 5, 6 415
NM_030597.3 NP_085100.1 4, 5 386
GenBank mRNA AF204146.1 - 4, 5 357
AF204147.1 - 4, 5 357
AF204148.1 - 4, 5 357
AF204149.1 - 4, 5 357
AF204150.1 - 4, 5 357
AF204151.1 - 4, 5 357
AF204152.1 - 4, 5 357
AF204153.1 - 3, 4 599
AF204154.1 - 5, 6 386
AF204155.1 - 5, 6 386
AF204156.1 - 4, 5 436
AF204157.1 - 4, 5 224
AK151605.1 - 4, 5 354
AK178855.1 - 4, 5 168
BB848823.1 - 4, 5 386
BC014288.1 - 4, 5 365 3982410
BC049543.1 - 379 6774564
BY079930.1 - 3, 4 314

View Full Product Details