Gene Transcripts

Gene Symbol
Dnal4
Entrez Gene ID
54152
Gene Name
dynein, axonemal, light chain 4
Gene Aliases
D15Ertd424e, Dnalc4
Chromosome Location
Chr. 15: 79761449 - 79774469 on Build GRCm38
UniGene ID
Mm.288159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017470.2 NP_059498.2 2 261
GenBank mRNA AB010031.1 - 2 239
AK002657.1 - 2 212
AK029618.1 - 1 72
AK132061.1 - 2 251
BC005426.1 - 2 190 3593932
BY012683.1 - 2 262

View Full Product Details