Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023455.3 | NP_075944.1 | 2 | 299 | |
GenBank mRNA | AF163317.1 | - | 2 | 256 | |
AK002784.1 | - | 2 | 240 | ||
BB846150.1 | - | 2 | 301 | ||
BC019517.1 | - | 2 | 491 | 4236543 | |
BY081766.1 | - | 2 | 280 |