Gene Transcripts

Gene Symbol
Prpf31
Entrez Gene ID
68988
Gene Name
PRP31 pre-mRNA processing factor 31 homolog (yeast)
Gene Aliases
1500019O16Rik, 2810404O06Rik, AW554706, PRP31, RP11
Chromosome Location
Chr. 7: 3629985 - 3642485 on Build GRCm38
UniGene ID
Mm.246863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159714.1 NP_001153186.1 8 814
NM_027328.4 NP_081604.3 8 814
GenBank mRNA AK005294.1 - 8 782
AK033190.1 - 8 817
AK033283.1 - 8 779
AK044457.1 - 7 2215
AK051260.1 - 8 774
AY040823.1 - 8 790
BC018376.1 - 8 780 3673893
BC057877.1 - 7 757 5347609
BC061461.1 - 8 758 5717606
CT010189.1 - 7 734

View Full Product Details