Gene Transcripts

Gene Symbol
Shisa4
Entrez Gene ID
77552
Gene Name
shisa family member 4
Gene Aliases
9330132O05Rik, Tmem58, mShisa4
Chromosome Location
Chr. 1: 135371057 - 135375277 on Build GRCm38
UniGene ID
Mm.41407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175259.4 NP_780468.1 3 437
GenBank mRNA AK039457.1 - 3 448
AK134719.1 - 1 220
BC044813.1 - 3 437 4950177

View Full Product Details