Gene Transcripts

Gene Symbol
Atp2b2
Entrez Gene ID
24215
Gene Name
ATPase, Ca++ transporting, plasma membrane 2
Gene Aliases
PMCA2
Chromosome Location
Chr. 4: 145704779 - 145948997 on Build Rnor_6.0
UniGene ID
Rn.90982
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012508.5 NP_036640.1 2984
GenBank mRNA J03754.1 - 2990

View Full Product Details