Gene Transcripts

Gene Symbol
Tspyl2
Entrez Gene ID
302612
Gene Name
TSPY-like 2
Gene Aliases
RGD1563283
Chromosome Location
Chr. X: 22408118 - 22413879 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191618.2 NP_001178547.1 5 1269
XM_006257038.2 XP_006257100.1 5 1420

View Full Product Details