Gene Transcripts

Gene Symbol
Map9
Entrez Gene ID
310544
Gene Name
microtubule-associated protein 9
Gene Aliases
Mtap9, RGD1560368
Chromosome Location
Chr. 2: 181331438 - 181368087 on Build Rnor_6.0
UniGene ID
Rn.198597
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135716.1 NP_001129188.1 14 2063
XM_006232515.2 XP_006232577.1 14 2125
XM_006232516.2 XP_006232578.1 13 1977

View Full Product Details