Gene Transcripts

Gene Symbol
Tanc1
Entrez Gene ID
311055
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases
Tanc
Chromosome Location
Chr. 3: 45718581 - 45917687 on Build Rnor_6.0
UniGene ID
Rn.161791
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002854.1 NP_001002854.1 20 3506
GenBank mRNA AB098072.1 - 20 3506

View Full Product Details