Gene Transcripts

Gene Symbol
Fcar
Entrez Gene ID
365183
Gene Name
Fc fragment of IgA, receptor for
Gene Aliases
CD89
Chromosome Location
Chr. 1: 73230392 - 73263462 on Build Rnor_6.0
UniGene ID
Rn.123516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201992.1 NP_973721.1 318
GenBank mRNA AB109766.1 - 318
AB109767.1 - 318
AB109768.1 - 274

View Full Product Details