Gene Transcripts

Gene Symbol
Psgb1
Entrez Gene ID
59313
Gene Name
pregnancy-specific beta 1-glycoprotein
Gene Aliases
Psg
Chromosome Location
Chr. 1: 79181855 - 79190738 on Build Rnor_6.0
UniGene ID
Rn.42878
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021677.1 NP_067709.1 3, 4 542
XM_006228464.1 XP_006228526.1 2, 3 411
GenBank mRNA U66292.1 - 3, 4 542

View Full Product Details