Gene Transcripts

Gene Symbol
Oas1d
Entrez Gene ID
304508
Gene Name
2'-5' oligoadenylate synthetase 1D
Gene Aliases
Oas1c
Chromosome Location
Chr. 12: 41179477 - 41188796 on Build Rnor_6.0
UniGene ID
Rn.110155
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009379.1 NP_001009379.1 1034
GenBank mRNA AY196699.1 - 1034

Gene Symbol
LOC103689988
Entrez Gene ID
103689988
Gene Name
2'-5'-oligoadenylate synthase 1A-like
Gene Aliases
Chromosome Location
Chr. 12: 42367110 - 42376531 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769299.1 XP_008767521.1 5 1134

View Full Product Details