Gene Transcripts

Gene Symbol
Llgl1
Entrez Gene ID
54265
Gene Name
lethal giant larvae homolog 1 (Drosophila)
Gene Aliases
LLGL, Llglh, Mgl1, Rgl-1, Rgl1
Chromosome Location
Chr. 10: 46940865 - 46955521 on Build Rnor_6.0
UniGene ID
Rn.122316
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246518.2 XP_006246580.1 12 1590
XM_006246519.2 XP_006246581.1 12 1587
GenBank mRNA AF356187.1 - 12 1435

View Full Product Details