Gene Transcripts
- Gene Symbol
- Enoph1
- Entrez Gene ID
- 67870
- Gene Name
- enolase-phosphatase 1
- Gene Aliases
- 2310057D15Rik, BB183658, C81437
- Chromosome Location
- Chr. 5: 100039994 - 100068925 on Build GRCm38
- UniGene ID
- Mm.11311
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027990.1 |
|
1, 2 |
278 |
|
GenBank mRNA |
CX235072.1 |
- |
1, 2 |
225 |
|
View Full Product Details