Gene Transcripts

Gene Symbol
Lypd6
Entrez Gene ID
320343
Gene Name
LY6/PLAUR domain containing 6
Gene Aliases
E130115E03Rik
Chromosome Location
Chr. 2: 50066429 - 50193569 on Build GRCm38
UniGene ID
Mm.130607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033304.2 2 279
GenBank mRNA AK053620.1 - 2 245
BC070462.1 - 1 135 30650880

View Full Product Details