Gene Transcripts
- Gene Symbol
- PRORSD1P
- Entrez Gene ID
- 344405
- Gene Name
- prolyl-tRNA synthetase associated domain containing 1, pseudogene
- Gene Aliases
- NCRNA00117, PRDXDD1P, Prdxdd1, Ybakd1
- Chromosome Location
- Chr. 2: 55282319 - 55284472 on Build GRCh38
- UniGene ID
- Hs.445292
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027258.1 |
|
1 |
1800 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE673666.1 |
- |
1 |
247 |
|
Chr. 2 - 55283898 - 55284407 [+] |
View Full Product Details