Gene Transcripts
- Gene Symbol
- PTPRVP
- Entrez Gene ID
- 148713
- Gene Name
- protein tyrosine phosphatase, receptor type V, pseudogene
- Gene Aliases
- ESP, OST-PTP, PTPRV
- Chromosome Location
- Chr. 1: 202168051 - 202189449 on Build GRCh38
- UniGene ID
- Hs.523870
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002930.2 |
|
6 |
1514 |
|
View Full Product Details