Gene Transcripts

Gene Symbol
TDH
Entrez Gene ID
157739
Gene Name
L-threonine dehydrogenase (pseudogene)
Gene Aliases
SDR14E1P
Chromosome Location
Chr. 8: 11339637 - 11368452 on Build GRCh38
UniGene ID
Hs.583896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001578.1 4 341
GenBank mRNA AJ301562.1 - 4 341
AK057762.1 - 5 430
AY101186.1 - 3 243
AY101187.1 - 3 243
BC074983.2 - 4 278


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX091784.1 - 4 296 Chr. 8 - 11346178 - 11361742 [+]

View Full Product Details