Gene Transcripts
- Gene Symbol
- SNORA69
- Entrez Gene ID
- 26779
- Gene Name
- small nucleolar RNA, H/ACA box 69
- Gene Aliases
- RNU69, U69, U69A
- Chromosome Location
- Chr. X: 119787353 - 119787484 on Build GRCh38
- UniGene ID
- Hs.675255
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002584.1 |
|
1 |
20 |
|
GenBank mRNA |
Y11163.1 |
- |
1 |
20 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX421532.2 |
- |
1 |
422 |
|
Chr. X - 119787067 - 119787886 [-] |
View Full Product Details