Gene Transcripts

Gene Symbol
KGFLP1
Entrez Gene ID
387628
Gene Name
fibroblast growth factor 7 pseudogene
Gene Aliases
Chromosome Location
Chr. 9: 62376256 - 62435519 on Build GRCh38
UniGene ID
Hs.439341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003674.2 3 1937
NR_047527.1 4 2047
GenBank mRNA AY098593.1 - 4 1868
CN274734.1 - 1 367

View Full Product Details