Gene Transcripts
- Gene Symbol
- TRIM53AP
- Entrez Gene ID
- 642569
- Gene Name
- tripartite motif containing 53A, pseudogene
- Gene Aliases
- TRIM53, TRIM53P, TRIM80P
- Chromosome Location
- Chr. 11: 89993541 - 90002508 on Build GRCh38
- UniGene ID
- Hs.647272
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028346.1 |
|
2 |
316 |
|
View Full Product Details