Gene Transcripts
- Gene Symbol
- SNORA26
- Entrez Gene ID
- 677810
- Gene Name
- small nucleolar RNA, H/ACA box 26
- Gene Aliases
- HBI-6
- Chromosome Location
- Chr. 4: 52713249 - 52713370 on Build GRCh38
- UniGene ID
- Hs.635034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003016.1 |
|
1 |
28 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM685854.1 |
- |
1 |
167 |
|
Chr. 4 - 52713116 - 52713681 [+] |
GenBankmRNA |
CN411930.1 |
- |
1 |
458 |
|
Chr. 4 - 52712823 - 52713547 [+] |
View Full Product Details