Gene Transcripts

Gene Symbol
SNORA80E
Entrez Gene ID
677823
Gene Name
small nucleolar RNA, H/ACA box 80E
Gene Aliases
ACA42, SNORA42
Chromosome Location
Chr. 1: 155919909 - 155920042 on Build GRCh38
UniGene ID
Hs.676501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002974.1 1 68
GenBank mRNA AJ609465.1 - 1 68


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU663871.1 - 83 Chr. - - [-]
GenBankmRNA DW418843.1 - 1 88 Chr. 1 - 155918992 - 155920049 [-]
GenBankmRNA DW454448.1 - 1 89 Chr. 1 - 155919883 - 155920049 [-]

View Full Product Details