Gene Transcripts
- Gene Symbol
- SNORA41
- Entrez Gene ID
- 619569
- Gene Name
- small nucleolar RNA, H/ACA box 41
- Gene Aliases
- ACA41, SNORA41A
- Chromosome Location
- Chr. 2: 206162228 - 206162359 on Build GRCh38
- UniGene ID
- Hs.742407
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002590.1 |
|
1 |
106 |
|
GenBank mRNA |
AJ609464.1 |
- |
1 |
106 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CV573603.1 |
- |
1 |
89 |
|
Chr. 2 - 206162245 - 206162904 [+] |
GenBankmRNA |
CA866694.1 |
- |
1 |
457 |
|
Chr. 2 - 206161888 - 206162522 [+] |
View Full Product Details