Gene Transcripts
- Gene Symbol
- SNORA15
- Entrez Gene ID
- 677803
- Gene Name
- small nucleolar RNA, H/ACA box 15
- Gene Aliases
- ACA15, SNORA15A
- Chromosome Location
- Chr. 7: 56060470 - 56060602 on Build GRCh38
- UniGene ID
- Hs.689706
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002957.1 |
|
1 |
25 |
|
GenBank mRNA |
AJ609458.1 |
- |
1 |
25 |
|
View Full Product Details