Gene Transcripts

Gene Symbol
BCAR4
Entrez Gene ID
400500
Gene Name
breast cancer anti-estrogen resistance 4 (non-protein coding)
Gene Aliases
Chromosome Location
Chr. 16: 11819830 - 11828832 on Build GRCh38
UniGene ID
Hs.24611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024049.1 1 473
NR_024050.1 2 166
NR_131217.1 2 381
GenBank mRNA BG573316.1 - 2 381
BX107175.1 - 2 137
BX336046.2 - 1 453

View Full Product Details