Gene Transcripts
- Gene Symbol
- SNORD8
- Entrez Gene ID
- 319103
- Gene Name
- small nucleolar RNA, C/D box 8
- Gene Aliases
- RNU6C, mgU6-53
- Chromosome Location
- Chr. 14: 21397293 - 21397401 on Build GRCh38
- UniGene ID
- Hs.680078
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002916.2 |
|
1 |
88 |
|
GenBank mRNA |
AJ243222.1 |
- |
1 |
87 |
|
View Full Product Details