Gene Transcripts

Gene Symbol
PRAC2
Entrez Gene ID
360205
Gene Name
prostate cancer susceptibility candidate 2
Gene Aliases
C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253
Chromosome Location
Chr. 17: 48720802 - 48724758 on Build GRCh38
UniGene ID
Hs.236557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282275.1 NP_001269204.1 2 158
NM_001282276.1 NP_001269205.1 2 92
XM_006721865.3 XP_006721928.1 2 774
XM_011524747.2 XP_011523049.1 2 84
XM_011524748.1 XP_011523050.1 2 29
GenBank mRNA AF532777.1 - 2 146
BX106051.1 - 2 92

Gene Symbol
LOC102724665
Entrez Gene ID
102724665
Gene Name
putative protein PRCA2
Gene Aliases
Chromosome Location
Chr. 6: 92644258 - 92645174 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017011600.1 XP_016867089.1 1 282


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB048981.1 - 2 122 Chr. 17 - 48723209 - 48724729 [+]

View Full Product Details