Gene Transcripts
- Gene Symbol
- TMCO5B
- Entrez Gene ID
- 100652857
- Gene Name
- transmembrane and coiled-coil domains 5B (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 15: 33236476 - 33247555 on Build GRCh38
- UniGene ID
- Hs.439998
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098530.1 |
- |
3 |
756 |
|
View Full Product Details