Gene Transcripts

Gene Symbol
DCAF12
Entrez Gene ID
25853
Gene Name
DDB1 and CUL4 associated factor 12
Gene Aliases
CT102, KIAA1892, TCC52, WDR40A
Chromosome Location
Chr. 9: 34086383 - 34126773 on Build GRCh38
UniGene ID
Hs.493750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF085907.1 - 1 58


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA R98118.1 - 53 Chr. - - [-]

View Full Product Details