Gene Transcripts

Gene Symbol
IRS4
Entrez Gene ID
8471
Gene Name
insulin receptor substrate 4
Gene Aliases
IRS-4, PY160
Chromosome Location
Chr. X: 108719949 - 108736584 on Build GRCh38
UniGene ID
Hs.407141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005262220.2 XP_005262277.1 2 6071
XM_011531061.1 XP_011529363.1 3 6097
GenBank mRNA AL832136.1 - 1 4486

View Full Product Details