Gene Transcripts
- Gene Symbol
- SIRT2
- Entrez Gene ID
- 22933
- Gene Name
- sirtuin 2
- Gene Aliases
- SIR2, SIR2L, SIR2L2
- Chromosome Location
- Chr. 19: 38878555 - 38899862 on Build GRCh38
- UniGene ID
- Hs.466693
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094810.1 |
- |
1 |
1644 |
|
View Full Product Details