Gene Transcripts
- Gene Symbol
- DLEU1
- Entrez Gene ID
- 10301
- Gene Name
- deleted in lymphocytic leukemia 1 (non-protein coding)
- Gene Aliases
- BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6
- Chromosome Location
- Chr. 13: 50082169 - 50528643 on Build GRCh38
- UniGene ID
- Hs.591229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ412029.1 |
- |
LIT1923 BCMS |
6 |
570 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AJ412022.3 |
- |
1 |
63 |
|
Chr. 13 - 50723522 - 50724063 [+] |
View Full Product Details