Gene Transcripts
- Gene Symbol
- NFE4
- Entrez Gene ID
- 58160
- Gene Name
- nuclear factor, erythroid 4
- Gene Aliases
- NF-E4
- Chromosome Location
- Chr. 7: 102973437 - 102978667 on Build GRCh38
- UniGene ID
- Hs.621233
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036938.1 |
- |
3 |
550 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI868837.1 |
- |
3 |
538 |
|
Chr. 7 - 102973523 - 102988608 [+] |
View Full Product Details