Gene Transcripts
- Gene Symbol
- Plch1
- Entrez Gene ID
- 269437
- Gene Name
- phospholipase C, eta 1
- Gene Aliases
- BC042549, PLCeta1, Plcl3
- Chromosome Location
- Chr. 3: 63696232 - 63899677 on Build GRCm38
- UniGene ID
- Mm.316391
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC048720.1 |
- |
2 |
1101 |
|
View Full Product Details