Gene Transcripts

Gene Symbol
Prickle2
Entrez Gene ID
243548
Gene Name
prickle homolog 2 (Drosophila)
Gene Aliases
6230400G14Rik, 6720451F06Rik, mpk2
Chromosome Location
Chr. 6: 92370891 - 92706335 on Build GRCm38
UniGene ID
Mm.248820
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020123.1 - 1 717


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DV063972.1 - 84 Chr. - - [-]

View Full Product Details